Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bex6
Entrez Gene ID
328660
Gene Name
brain expressed gene 6
Gene Aliases
B020003O03Rik
Chromosome Location
Chr.16: 32179800 - 32186944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033539.2 NP_001028711.1 3 - 3 539 81
GenBank mRNA AK139543.1 - 2 - 2 470 81
AK157452.1 - 3 - 3 540 81
AK163328.1 - 2 - 2 422 81
AY833559.1 - 3 - 3 539 81
BC132130.1 - 1 - 1 309 81
BC132132.1 - 1 - 1 309 81

View Full Product Details