Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Armc10
Entrez Gene ID
67211
Gene Name
armadillo repeat containing 10
Gene Aliases
2810037C14Rik, Svh
Chromosome Location
Chr.5: 21645984 - 21662592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026034.4 NP_080310.1 1 - 2 255 155
GenBank mRNA AK009821.1 - 1 - 2 248 155
AK011303.1 - 1 - 2 254 155
AK012872.1 - 1 - 2 230 155
BC027343.1 - 1 - 2 186 155
BC038487.1 - 1 - 2 207 155
BC046281.1 - 1 - 2 207 155
BC058573.1 - 1 - 2 296 155

View Full Product Details