The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026034.4 | NP_080310.1 | 5 - 6 | 784 | 81 |
GenBank mRNA | AK009821.1 | - | 5 - 6 | 778 | 81 |
AK011303.1 | - | 5 - 6 | 783 | 81 | |
AK012872.1 | - | 5 - 6 | 759 | 81 | |
AK015272.1 | - | 5 - 6 | 917 | 81 | |
BC038487.1 | - | 5 - 6 | 736 | 81 | |
BC046281.1 | - | 5 - 6 | 736 | 81 | |
BC058573.1 | - | 5 - 6 | 825 | 81 |