Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfwd3
Entrez Gene ID
234736
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
BC027246
Chromosome Location
Chr.8: 111270944 - 111300260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146218.4 NP_666330.2 5 - 6 1031 91
GenBank mRNA AK137499.1 - 5 - 6 1011 91
AK137698.1 - 5 - 6 994 91
AK139032.1 - 5 - 6 1012 91
AK146451.1 - 5 - 6 1001 91
BC018533.1 - 5 - 6 1000 91
BC023694.1 - 5 - 6 1036 91
BC096602.1 - 5 - 6 994 91

View Full Product Details