Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esp1
Entrez Gene ID
619517
Gene Name
exocrine gland secreted peptide 1
Gene Aliases
EG619517, Gm6084
Chromosome Location
Chr.17: 40727120 - 40731782 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038500.2 NP_001033589.1 2 - 3 159 122
GenBank mRNA AB194091.1 - 2 - 3 188 122

View Full Product Details