Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinb6e
Entrez Gene ID
435350
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 6e
Gene Aliases
Gm11396, OTTMUSG00000000720, SPI3B, ovalbumin
Chromosome Location
Chr.13: 33832292 - 33843408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045535.2 NP_001039000.2 4 - 5 559 130

View Full Product Details