Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Olfr55
- Entrez Gene ID
-
100038859
- Gene Name
- olfactory receptor 55
- Gene Aliases
-
IF6, MOR267-17, MOR267-20
- Chromosome Location
-
Chr.17: 33176416 - 33177363 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010998.2 |
NP_035128.2 |
1 - 1 |
836 |
146 |
- Gene Symbol
- Olfr239
- Entrez Gene ID
-
100038860
- Gene Name
- olfactory receptor 239
- Gene Aliases
-
ENSMUSG00000079308, MOR267-10
- Chromosome Location
-
Chr.17: 33199062 - 33200009 on Build GRCm38
View Full Product Details