Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdf3
Entrez Gene ID
14562
Gene Name
growth differentiation factor 3
Gene Aliases
C78318, Gdf-3, Vgr-2, Vgr2, ecat9
Chromosome Location
Chr.6: 122605403 - 122610071 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008108.4 NP_032134.2 1 - 2 376 178
GenBank mRNA AK082877.1 - 1 - 2 297 178
AK146174.1 - 1 - 2 302 178
AK152457.1 - 1 - 2 377 178
AK160533.1 - 1 - 2 334 178
AK167962.1 - 1 - 2 302 178
BC101963.1 - 1 - 2 291 178
BC101964.1 - 1 - 2 291 178
BC101965.1 - 1 - 2 179
BC103565.1 - 1 - 2 290 178
L06443.1 - 1 - 2 392 178
S52658.1 - 1 - 2 332 178

View Full Product Details