The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008540.2 | NP_032566.2 | 3 - 4 | 861 | 71 |
GenBank mRNA | AB221585.1 | - | 2 - 3 | 421 | 71 |
AK004804.1 | - | 2 - 3 | 225 | 71 | |
AY493561.1 | - | 2 - 3 | 421 | 71 | |
BC046584.1 | - | 3 - 4 | 861 | 71 | |
GU072917.1 | - | 2 - 3 | 421 | 71 | |
GU072918.1 | - | 2 - 3 | 421 | 71 | |
GU072919.1 | - | 2 - 3 | 421 | 71 | |
GU072920.1 | - | 2 - 3 | 421 | 71 | |
U79748.1 | - | 2 - 3 | 741 | 71 |