Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lefty2
Entrez Gene ID
320202
Gene Name
left-right determination factor 2
Gene Aliases
6030463A22Rik, AV214969, Ebaf, Lefta, Leftb
Chromosome Location
Chr.1: 180893119 - 180899103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177099.3 NP_796073.1 2 - 2 448 154
GenBank mRNA AK031621.1 - 2 - 2 460 154
AK131960.1 - 2 - 2 461 154
BC066224.1 - 2 - 2 448 154

View Full Product Details