The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011471.2 | NP_035601.1 | 2 - 2 | 447 | 123 |
GenBank mRNA | AY158989.1 | - | 2 - 2 | 447 | 123 |