The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025596.5 | NP_079872.4 | 5 - 5 | 949 | 149 |
GenBank mRNA | AK002846.1 | - | 5 - 5 | 973 | 149 |
AK003629.1 | - | 4 - 4 | 693 | 149 | |
AK146857.1 | - | 5 - 5 | 979 | 149 | |
AK150465.1 | - | 5 - 5 | 968 | 149 | |
AK151321.1 | - | 5 - 5 | 968 | 149 | |
AK153455.1 | - | 5 - 5 | 973 | 149 | |
AK155042.1 | - | 5 - 5 | 966 | 149 | |
BC024813.1 | - | 5 - 5 | 747 | 149 | |
BC025859.1 | - | 5 - 5 | 949 | 149 | |
BC059038.1 | - | - | 2504 | 149 | |
BC098241.1 | - | 5 - 5 | 956 | 149 |