The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080387.2 | NP_001073856.1 | 1 - 2 | 224 | 117 |
NM_001284195.1 | NP_001271124.1 | 1 - 2 | 224 | 117 | |
NM_001284196.1 | NP_001271125.1 | 1 - 2 | 224 | 117 | |
NM_010178.3 | NP_034308.1 | 1 - 2 | 224 | 117 | |
GenBank mRNA | AB015894.1 | - | 1 - 2 | 135 | 117 |
AB015895.1 | - | 1 - 2 | 132 | 117 | |
AF042383.1 | - | 1 - 2 | 136 | 117 | |
AF060490.1 | - | 1 - 2 | 159 | 117 | |
AK005295.1 | - | 1 - 2 | 166 | 117 | |
AK014345.1 | - | 1 - 2 | 163 | 117 | |
AK134076.1 | - | 1 - 2 | 194 | 117 | |
AK134289.1 | - | 1 - 2 | 194 | 117 | |
AK135316.1 | - | 1 - 2 | 169 | 117 | |
AK149716.1 | - | 1 - 2 | 151 | 117 | |
AK150072.1 | - | 1 - 2 | 151 | 117 | |
AK150734.1 | - | 1 - 2 | 158 | 117 | |
AK151567.1 | - | 1 - 2 | 163 | 117 | |
AK161097.1 | - | 1 - 2 | 183 | 117 | |
AK168524.1 | - | 1 - 2 | 182 | 117 | |
AK168558.1 | - | 1 - 2 | 182 | 117 | |
AK168571.1 | - | 1 - 2 | 147 | 117 | |
AK169071.1 | - | 1 - 2 | 151 | 117 | |
BC006670.1 | - | 1 - 2 | 176 | 117 | |
BC043060.1 | - | 1 - 2 | 548 | 117 | |
BC083082.1 | - | 1 - 2 | 136 | 117 | |
BY717856.1 | - | 1 - 2 | 194 | 117 | |
CF914989.1 | - | 1 - 2 | 140 | 117 |