The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025575.2 | NP_001020746.1 | 4 - 5 | 685 | 145 |
GenBank mRNA | AJ565908.1 | - | 4 - 5 | 634 | 145 |
AK149511.1 | - | 4 - 5 | 676 | 145 | |
M29007.1 | - | - | 677 | 145 |