Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr12
Entrez Gene ID
57750
Gene Name
WD repeat domain 12
Gene Aliases
4933402C23Rik, Ytm1, Ytm1p
Chromosome Location
Chr.1: 60076868 - 60098500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199060.1 NP_001185989.1 11 - 12 1521 86
NM_001199061.1 NP_001185990.1 11 - 12 1253 86
NM_021312.5 NP_067287.1 10 - 11 1343 86
GenBank mRNA AB041608.1 - 11 - 12 1522 86
AF239765.1 - 10 - 11 999 86
AK007500.1 - 10 - 11 1120 86
AK016611.1 - 11 - 12 1197 86
AK031436.1 - 10 - 11 1106 86
AK149085.1 - 10 - 11 1025 86
AK161299.1 - 11 - 12 1266 86
AY059431.1 - 10 - 11 1034 86
AY059432.1 - 3 - 4 339 86
BC004748.1 - 10 - 11 1177 86
BC052386.1 - 10 - 11 1314 86
BC096651.1 - 10 - 11 1088 86

View Full Product Details