The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025860.3 | NP_080136.2 | 4 - 5 | 726 | 87 |
GenBank mRNA | AK019845.1 | - | 4 - 5 | 650 | 87 |
AK075864.1 | - | 4 - 5 | 663 | 87 | |
BC028246.1 | - | 4 - 5 | 697 | 87 | |
BC103776.1 | - | 4 - 5 | 663 | 87 | |
BC141230.1 | - | 4 - 5 | 665 | 87 | |
BC141231.1 | - | 4 - 5 | 665 | 87 |