The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130529.2 | NP_001124001.1 | 4 - 5 | 1100 | 80 |
NM_172941.4 | NP_766529.3 | 4 - 5 | 1147 | 80 | |
GenBank mRNA | AK139346.1 | - | 4 - 5 | 1390 | 80 |
AK155196.1 | - | 4 - 5 | 677 | 80 | |
AK155490.1 | - | 4 - 5 | 729 | 80 | |
AK158448.1 | - | 4 - 5 | 685 | 80 | |
AK170924.1 | - | 4 - 5 | 677 | 80 | |
AK170989.1 | - | 4 - 5 | 682 | 80 | |
AK171190.1 | - | 4 - 5 | 729 | 80 | |
AK178776.1 | - | 1 - 2 | 65 | 80 | |
AK211459.1 | - | 1 - 2 | 65 | 80 | |
AY302242.1 | - | 3 - 4 | 628 | 80 | |
BC040205.1 | - | 3 - 4 | 632 | 80 | |
CN530086.1 | - | 2 - 3 | 547 | 80 |