Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zscan2
Entrez Gene ID
22691
Gene Name
zinc finger and SCAN domain containing 2
Gene Aliases
Zfp-29, Zfp29
Chromosome Location
Chr.7: 80860737 - 80878128 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009553.2 NP_033579.1 2 - 3 709 91
GenBank mRNA AB221592.1 - 1 - 2 406 91
AK147779.1 - 2 - 3 605 91
BC046961.1 - 2 - 3 709 91
X55126.1 - 1 - 2 507 91

View Full Product Details