The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177794.1 | NP_001171265.1 | 3 - 4 | 359 | 67 |
NM_198247.2 | NP_937890.2 | 3 - 4 | 614 | 67 | |
GenBank mRNA | BC060251.1 | - | 3 - 4 | 583 | 67 |
BX514346.1 | - | 3 - 4 | 360 | 67 |