The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008960.2 | NP_032986.1 | 9 - 9 | 8156 | 91 |
GenBank mRNA | AK138025.1 | - | 2 - 2 | 6464 | 91 |
BU701098.1 | - | 1 - 1 | 91 |