The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026991.2 | NP_081267.1 | 5 - 6 | 504 | 121 |
GenBank mRNA | AK011406.1 | - | 6 - 7 | 423 | 121 |
BC061227.1 | - | 5 - 6 | 504 | 121 |