Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim34a
Entrez Gene ID
94094
Gene Name
tripartite motif-containing 34A
Gene Aliases
Trim34, Trim34-1
Chromosome Location
Chr.7: 104244437 - 104262239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030684.3 NP_109609.2 6 - 7 1202 129
GenBank mRNA AF220139.1 - 6 - 7 1099 129

View Full Product Details