Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r97
Entrez Gene ID
627367
Gene Name
vomeronasal 2, receptor 97
Gene Aliases
EG627367, Vn2r97
Chromosome Location
Chr.17: 18914322 - 18948071 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104549.1 NP_001098019.1 3 - 4 1344 150

View Full Product Details