The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027316.1 | 8 - 9 | 1766 | 103 | |
GenBank mRNA | AK138248.1 | - | 8 - 9 | 1766 | 103 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ribosomal RNA processing 1 homolog B (S. cerevisiae) | Rrp1b | 2600005C20Rik, D030064A17, Kiaa0179, mKIAA0179 | -13.2kb | Anti-sense | Chr. - 32036162 - 32062865 on Build GRCm38 |
Downstream | predicted gene, 38585 | Gm38585 | 36.9kb | Anti-sense | Chr. - 32113924 - 32129237 on Build GRCm38 |