Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srsf2
Entrez Gene ID
20382
Gene Name
serine/arginine-rich splicing factor 2
Gene Aliases
D11Wsu175e, MRF-1, Pr264, SC35, Sfrs10, Sfrs2
Chromosome Location
Chr.11: 116849897 - 116853104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011358.2 NP_035488.1 1 - 2 531 106
GenBank mRNA AF077858.1 - 1 - 2 519 106
AK086103.1 - 1 - 2 529 106
AK088042.1 - 1 - 2 529 106
AK161873.1 - 1 - 2 527 106
AK165670.1 - 1 - 2 533 106
BC005493.1 - 1 - 2 509 106
CJ043688.1 - 1 - 2 532 106
L13610.1 - 1 - 2 503 106

View Full Product Details