Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tead1
- Entrez Gene ID
-
21676
- Gene Name
- TEA domain family member 1
- Gene Aliases
-
2610024B07Rik, B230114H05Rik, Gtrgeo5, TEAD-1, TEF-1, Tcf13, mTEF-1
- Chromosome Location
-
Chr.7: 112679320 - 112906805 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CF907453.1 |
- |
1 |
275 |
|
Chr. 7 - 112903753 - 112904334 [+] on Build GRCm38 |
89 |
GenBankmRNA |
BU514882.1 |
- |
1 |
655 |
|
Chr. 7 - 112903373 - 112904202 [+] on Build GRCm38 |
89 |
GenBankmRNA |
CN698958.1 |
- |
1 |
276 |
|
Chr. 7 - 112903752 - 112904410 [+] on Build GRCm38 |
89 |
GenBankmRNA |
CA980385.1 |
- |
1 |
507 |
|
Chr. 7 - 112903521 - 112904346 [+] on Build GRCm38 |
89 |
GenBankmRNA |
CN532645.1 |
- |
1 |
90 |
|
Chr. 7 - 112903938 - 112904653 [+] on Build GRCm38 |
89 |
GenBankmRNA |
CF909223.1 |
- |
1 |
245 |
|
Chr. 7 - 112903783 - 112904310 [+] on Build GRCm38 |
89 |
GenBankmRNA |
AV579760.1 |
- |
1 |
75 |
|
Chr. 7 - 112903954 - 112904461 [+] on Build GRCm38 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 2310014F06 gene |
2310014F06Rik |
|
-239.1kb
|
Anti-sense |
Chr. - 112596533 - 112664189 on Build GRCm38 |
Downstream |
Ras association (RalGDS/AF-6) domain family (N-terminal) member 10 |
Rassf10 |
4632411J06Rik |
49.0kb
|
Sense |
Chr. - 112953962 - 112957458 on Build GRCm38 |
View Full Product Details