Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dennd1b
Entrez Gene ID
329260
Gene Name
DENN/MADD domain containing 1B
Gene Aliases
4632404N19Rik, 4930467M19Rik, 6820401H01Rik, F730008N07Rik
Chromosome Location
Chr.1: 138963446 - 139176043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166501.1 NP_001159973.1 22 - 22 6279 104
NM_181347.3 NP_851992.1 21 - 21 6145 104
GenBank mRNA AK018353.1 - 1 - 1 409 104
AK039998.1 - 1 - 1 448 103


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY719473.1 - 409 Chr. - - [-] on Build 104


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RIKEN cDNA 4930596I21 gene 4930596I21Rik -194.4kb Sense Chr. - 138977657 - 138979246 on Build GRCm38
Downstream crumbs family member 1, photoreceptor morphogenesis associated Crb1 7530426H14Rik, A930008G09Rik 22.3kb Anti-sense Chr. - 139197546 - 139379118 on Build GRCm38

View Full Product Details