Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apaf1
Entrez Gene ID
11783
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
6230400I06Rik, Apaf-1, Apaf1l, fog, mKIAA0413
Chromosome Location
Chr.10: 90989311 - 91082743 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK053432.1 - 1 - 1 3351 74


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU258882.1 - 1 149 Chr. 10 - 91038092 - 91038435 [-] on Build GRCm38 74
GenBankmRNA BB539201.2 - 1 286 Chr. 10 - 91037936 - 91038572 [-] on Build GRCm38 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ankyrin repeat and sterile alpha motif domain containing 1B Anks1b AIDA-1b, C030032C09Rik, E530015N03Rik, ENSMUSG00000078431, Gm10937, Gm1555 -64.6kb Anti-sense Chr. - 89873509 - 90973300 on Build GRCm38
Downstream IKBKB interacting protein Ikbip 1200009F10Rik, 1700023M03Rik, D18354, Ikip 41.4kb Anti-sense Chr. - 91083039 - 91102613 on Build GRCm38

View Full Product Details