Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gabra2
- Entrez Gene ID
-
14395
- Gene Name
- gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2
- Gene Aliases
-
C630048P16Rik, Gabra-2
- Chromosome Location
-
Chr.5: 70961037 - 71095849 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK036049.1 |
- |
- |
738 |
88 |
AK079593.1 |
- |
- |
944 |
88 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW124947.1 |
- |
1 |
|
|
Chr. 5 - 70959145 - 70959573 [+] on Build GRCm38 |
88 |
GenBankmRNA |
BQ174589.1 |
- |
1 |
|
|
Chr. 5 - 70959145 - 70959808 [+] on Build GRCm38 |
88 |
GenBankmRNA |
BU609553.1 |
- |
1 |
146 |
|
Chr. 5 - 70958825 - 70959592 [-] on Build GRCm38 |
88 |
GenBankmRNA |
CO424865.1 |
- |
1 |
160 |
|
Chr. 5 - 70958967 - 70959606 [-] on Build GRCm38 |
88 |
GenBankmRNA |
BE650342.1 |
- |
1 |
439 |
|
Chr. 5 - 70959368 - 70959885 [-] on Build GRCm38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
gamma-aminobutyric acid (GABA) A receptor, subunit gamma 1 |
Gabrg1 |
GabaA, GabaA/BZ |
-115.6kb
|
Anti-sense |
Chr. - 70751047 - 70843508 on Build GRCm38 |
View Full Product Details