The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK139201.1 | - | 1 - 2 | 179 | 89 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | cell adhesion molecule 1 | Cadm1 | 2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1 | -196.1kb | Sense | Chr. - 47530239 - 47853385 on Build GRCm38 |
Downstream | neurexophilin and PC-esterase domain family, member 4 | Nxpe4 | C130036J11, D930028F11Rik, Fam55d | 111.3kb | Sense | Chr. - 48162001 - 48400025 on Build GRCm38 |