The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010553.4 | NP_034683.1 | 13 - 13 | 4338 | 103 |
GenBank mRNA | AK161196.1 | - | 1 - 1 | 2246 | 103 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | interleukin 18 receptor 1 | Il18r1 | Il18ralpha, Il1rrp | -48.1kb | Sense | Chr. - 40465552 - 40500854 on Build GRCm38 |
Downstream | solute carrier family 9 (sodium/hydrogen exchanger), member 4 | Slc9a4 | AW990558, D730009J23Rik, NHE4 | 28.0kb | Sense | Chr. - 40579715 - 40630731 on Build GRCm38 |