Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mcu
- Entrez Gene ID
-
215999
- Gene Name
- mitochondrial calcium uniporter
- Gene Aliases
-
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
- Chromosome Location
-
Chr.10: 59446984 - 59616692 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK166044.1 |
- |
2 - 3 |
262 |
83 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY734336.1 |
- |
2 - 3 |
262 |
|
Chr. 10 - 59534799 - 59616675 [-] on Build GRCm38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
oncoprotein induced transcript 3 |
Oit3 |
EF-9, LZP |
-158.1kb
|
Sense |
Chr. - 59422960 - 59441779 on Build GRCm38 |
Downstream |
mitochondrial calcium uptake 1 |
Micu1 |
C730016L05Rik, Calc, Cbara1 |
101.8kb
|
Anti-sense |
Chr. - 59702482 - 59864134 on Build GRCm38 |
View Full Product Details