The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_001463.3 | 7 - 7 | 14131 | 102 | |
GenBank mRNA | AK011511.1 | - | 1 - 1 | 245 | 102 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | testis specific X-linked gene | Tsx | AV207184 | -38.9kb | Anti-sense | Chr. - 103409101 - 103424588 on Build GRCm38 |
Downstream | Jpx transcript, Xist activator (non-protein coding) | Jpx | 2010000I03Rik, 2510040I06Rik, Enox | 29.2kb | Anti-sense | Chr. - 103493576 - 103506425 on Build GRCm38 |