The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | Y17853.1 | - | 1 - 1 | 52 | 93 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | replication protein A1 | Rpa1 | 5031405K23Rik, 70kDa, AA589576, AW557552, RF-A, RP-A, Rpa | -35.6kb | Anti-sense | Chr. - 75300259 - 75348383 on Build GRCm38 |
Downstream | serine (or cysteine) peptidase inhibitor, clade F, member 1 | Serpinf1 | AI195227, EPC-1, Pedf, Pedfl, Sdf3 | 25.8kb | Anti-sense | Chr. - 75410029 - 75422623 on Build GRCm38 |