Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK044545.1 - 49 Chr. - - [-] on Build 97
GenBankmRNA BB642537.1 - 1 49 Chr. 9 - 107680082 - 107680742 [+] on Build GRCm38 97
GenBankmRNA BG295830.1 - 1 241 Chr. 9 - 107679894 - 107680649 [+] on Build GRCm38 97
GenBankmRNA BI734089.1 - 1 273 Chr. 9 - 107679856 - 107680737 [+] on Build GRCm38 97
GenBankmRNA BU506259.1 - 1 274 Chr. 9 - 107679856 - 107680766 [+] on Build GRCm38 97


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream guanine nucleotide binding protein, alpha transducing 1 Gnat1 Gnat-1, Ird1, Ird2, Tralpha, irdc, irdr, transducin -264.0 Anti-sense Chr. - 107674474 - 107679592 on Build GRCm38
Downstream sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F Sema3f Sema4, Semak 733.0 Anti-sense Chr. - 107681499 - 107710475 on Build GRCm38

View Full Product Details