Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Enah
Entrez Gene ID
13800
Gene Name
enabled homolog (Drosophila)
Gene Aliases
Mena, NDPP-1, Ndpp1, WBP8
Chromosome Location
Chr.1: 181896384 - 182019980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083120.2 NP_001076589.1 14 - 14 8631 86
NM_001083121.2 NP_001076590.1 14 - 14 7899 86
NM_008680.3 NP_032706.2 15 - 15 8643 86
NM_010135.3 NP_034265.2 15 - 15 8688 86
GenBank mRNA AK081526.1 - 1 - 1 1853 86


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB519783.1 - 1 498 Chr. 1 - 181899776 - 181900530 [-] on Build GRCm38 86
GenBankmRNA AW121512.1 - 1 Chr. 1 - 181899793 - 181900360 [+] on Build GRCm38 86
GenBankmRNA CK639844.1 - 1 196 Chr. 1 - 181899451 - 181900228 [-] on Build GRCm38 86
GenBankmRNA CB244257.1 - 1 491 Chr. 1 - 181899821 - 181900523 [-] on Build GRCm38 86


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream lamin B receptor Lbr AI505894, ic -57.5kb Sense Chr. - 181815315 - 181842401 on Build GRCm38
Downstream signal recognition particle 9 Srp9 9kDa 222.9kb Anti-sense Chr. - 182124737 - 182132415 on Build GRCm38

View Full Product Details