Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB480463.2 - 1 489 Chr. 7 - 143350424 - 143351050 [-] on Build GRCm38 97
GenBankmRNA AK084816.1 - 3352 Chr. - - [-] on Build 97


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream potassium voltage-gated channel, subfamily Q, member 1 Kcnq1 AW559127, KVLQT1, Kcna9 Anti-sense Chr. - 143106742 - 143427050 on Build GRCm38
Downstream cyclin-dependent kinase inhibitor 1C (P57) Cdkn1c AL024410, CDKI, Kip2, p57(kip2), p57Kip2 107.3kb Sense Chr. - 143458339 - 143461072 on Build GRCm38

View Full Product Details