Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gm15958
Entrez Gene ID
100503546
Gene Name
predicted gene 15958
Gene Aliases
OTTMUSG00000028223
Chromosome Location
Chr.18: 66312259 - 66319608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK138348.1 - - 453 92


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY724018.1 - 2 - 3 453 Chr. 18 - 66291274 - 66317007 [+] on Build GRCm38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream collagen and calcium binding EGF domains 1 Ccbe1 4933426F18Rik, 9430093N24Rik, mKIAA1983 -20.4kb Anti-sense Chr. - 66056855 - 66291838 on Build GRCm38
Downstream phorbol-12-myristate-13-acetate-induced protein 1 Pmaip1 Noxa 141.6kb Sense Chr. - 66458604 - 66465558 on Build GRCm38

View Full Product Details