Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG082444.2 - 1 125 Chr. 11 - 74680178 - 74680682 [-] on Build GRCm38 92
GenBankmRNA AK162339.1 - 2345 Chr. - - [-] on Build 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream platelet-activating factor acetylhydrolase, isoform 1b, subunit 1 Pafah1b1 LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha Sense Chr. - 74673949 - 74724384 on Build GRCm38
Downstream methyltransferase like 16 Mettl16 2610100D03Rik, 2810013M15Rik, A830095F14Rik, AI846133, Mett10d 90.2kb Anti-sense Chr. - 74770836 - 74818694 on Build GRCm38

View Full Product Details