Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ND4L
Entrez Gene ID
17720
Gene Name
NADH dehydrogenase subunit 4L
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK140187.1 - 1 - 1 345 81
AK153747.1 - - 344 81
AK157367.1 - - 345 81

View Full Product Details