Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap5-3
Entrez Gene ID
77215
Gene Name
keratin associated protein 5-3
Gene Aliases
A030007E19Rik
Chromosome Location
Chr.7: 142201364 - 142203015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023860.1 NP_076349.1 1 - 1 1449 85
GenBank mRNA AK020699.1 - 2 - 2 872 85

View Full Product Details