The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027198.1 | NP_081474.1 | 4 - 5 | 367 | 96 |
GenBank mRNA | AK010472.1 | - | 4 - 5 | 347 | 96 |
BC109343.2 | - | 4 - 5 | 325 | 96 | |
BQ554850.1 | - | 4 - 5 | 370 | 96 |