Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap2-4
Entrez Gene ID
71453
Gene Name
keratin associated protein 2-4
Gene Aliases
5530401P19Rik, KAP2.4, KRTAP2.4
Chromosome Location
Chr.11: 99614017 - 99614846 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027800.1 NP_082076.1 1 - 1 752 84
GenBank mRNA AI892716.1 - 1 - 1 90
AK017442.1 - 1 - 1 752 84

View Full Product Details