Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zscan29
Entrez Gene ID
99334
Gene Name
zinc finger SCAN domains 29
Gene Aliases
AW125441, Zfp690
Chromosome Location
Chr.2: 121158273 - 121171125 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290819.1 NP_001277748.1 4 - 5 2653 97

View Full Product Details