The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048060.2 | NP_001041525.1 | 1 - 2 | 92 | 86 |
NM_198004.3 | NP_932121.2 | 1 - 2 | 92 | 86 | |
GenBank mRNA | AK017224.1 | - | 1 - 2 | 87 | 86 |
AK032811.1 | - | 1 - 2 | 75 | 86 | |
AK040588.1 | - | 1 - 2 | 93 | 86 | |
AK045763.1 | - | 1 - 2 | 88 | 86 | |
BC026742.1 | - | 1 - 2 | 61 | 86 | |
BU525297.1 | - | 1 - 2 | 95 | 86 | |
CX206109.1 | - | 1 - 2 | 68 | 86 |