Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfpl
Entrez Gene ID
56093
Gene Name
pore forming protein-like
Gene Aliases
Epcs5, Epcs50
Chromosome Location
Chr.19: 12427905 - 12432110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019540.2 NP_062413.2 1 - 2 144 104
GenBank mRNA AF250839.1 - 1 - 2 120 104
AK141262.1 - 1 - 2 78 104
BC052215.1 - 1 - 2 112 104
CJ075286.1 - 1 - 2 144 104

View Full Product Details