Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm2083
- Entrez Gene ID
-
100048885
- Gene Name
- major urinary protein LOC100048885
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- LOC100048884
- Entrez Gene ID
-
100048884
- Gene Name
- novel member of the major urinary protein (Mup) gene family
- Gene Aliases
-
Mup6
- Chromosome Location
-
Chr.4: 61670177 - 61674098 on Build GRCm38
- Gene Symbol
- Mup1
- Entrez Gene ID
-
17840
- Gene Name
- major urinary protein 1
- Gene Aliases
-
Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
- Chromosome Location
-
Chr.4: 60498012 - 60501960 on Build GRCm38
- Gene Symbol
- Mup10
- Entrez Gene ID
-
100039008
- Gene Name
- major urinary protein 10
- Gene Aliases
-
2610016E04Rik, Mup8
- Chromosome Location
-
Chr.4: 60578260 - 60582155 on Build GRCm38
- Gene Symbol
- Mup11
- Entrez Gene ID
-
100039028
- Gene Name
- major urinary protein 11
- Gene Aliases
-
Gm12549, OTTMUSG00000007431
- Chromosome Location
-
Chr.4: 60658466 - 60662395 on Build GRCm38
- Gene Symbol
- Mup12
- Entrez Gene ID
-
100039054
- Gene Name
- major urinary protein 12
- Gene Aliases
-
Gm2024
- Chromosome Location
-
Chr.4: 60737381 - 60741281 on Build GRCm38
- Gene Symbol
- Mup13
- Entrez Gene ID
-
100039089
- Gene Name
- major urinary protein 13
- Gene Aliases
-
Gm13513, Mup11, OTTMUSG00000012492
- Chromosome Location
-
Chr.4: 61224307 - 61228286 on Build GRCm38
- Gene Symbol
- Mup15
- Entrez Gene ID
-
100039150
- Gene Name
- major urinary protein 15
- Gene Aliases
-
Gm2068, Mup13
- Chromosome Location
-
Chr.4: 61435787 - 61439714 on Build GRCm38
- Gene Symbol
- Mup16
- Entrez Gene ID
-
100039177
- Gene Name
- major urinary protein 16
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 61515592 - 61519503 on Build GRCm38
- Gene Symbol
- Mup17
- Entrez Gene ID
-
100039206
- Gene Name
- major urinary protein 17
- Gene Aliases
-
Gm12557, MUP 17, OTTMUSG00000007480
- Chromosome Location
-
Chr.4: 61591929 - 61595853 on Build GRCm38
- Gene Symbol
- Mup19
- Entrez Gene ID
-
100189605
- Gene Name
- major urinary protein 19
- Gene Aliases
-
100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
- Chromosome Location
-
Chr.4: 61778324 - 61782236 on Build GRCm38
- Gene Symbol
- Mup2
- Entrez Gene ID
-
17841
- Gene Name
- major urinary protein 2
- Gene Aliases
-
AA589603, Mup-2, Mup4
- Chromosome Location
-
Chr.4: 60135913 - 60139872 on Build GRCm38
- Gene Symbol
- Mup7
- Entrez Gene ID
-
100041658
- Gene Name
- major urinary protein 7
- Gene Aliases
-
Gm12546, Mup3, OTTMUSG00000007428
- Chromosome Location
-
Chr.4: 60066469 - 60070481 on Build GRCm38
- Gene Symbol
- Mup9
- Entrez Gene ID
-
100038948
- Gene Name
- major urinary protein 9
- Gene Aliases
-
Gm14076, Mup2, Mup6, OTTMUSG00000015595
- Chromosome Location
-
Chr.4: 60418046 - 60421961 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC094452.1 |
- |
|
|
|
Chr. - - [-] on Build |
104 |
View Full Product Details