The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048060.2 | NP_001041525.1 | 2 - 3 | 119 | 83 |
NM_198004.3 | NP_932121.2 | 2 - 3 | 119 | 83 | |
GenBank mRNA | AK017224.1 | - | 2 - 3 | 114 | 83 |
AK032811.1 | - | 2 - 3 | 102 | 83 | |
AK040588.1 | - | 2 - 3 | 120 | 83 | |
AK045763.1 | - | 2 - 3 | 115 | 83 | |
BC026742.1 | - | 2 - 3 | 88 | 83 | |
BU525297.1 | - | 2 - 3 | 122 | 83 | |
CX206109.1 | - | 2 - 3 | 95 | 83 |