The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081140.1 | NP_001074609.1 | 1 - 2 | 99 | 97 |
GenBank mRNA | AK157965.1 | - | 1 - 2 | 100 | 97 |
BC137828.1 | - | 1 - 2 | 188 | 97 | |
BC137829.1 | - | 1 - 2 | 188 | 97 |