Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r31
- Entrez Gene ID
-
100042591
- Gene Name
- vomeronasal 2, receptor 31
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 7383985 - 7399627 on Build GRCm38
- Gene Symbol
- Vmn2r39
- Entrez Gene ID
-
545909
- Gene Name
- vomeronasal 2, receptor 39
- Gene Aliases
-
EG545909
- Chromosome Location
-
Chr.7: 9014750 - 9030682 on Build GRCm38
- Gene Symbol
- Vmn2r43
- Entrez Gene ID
-
381838
- Gene Name
- vomeronasal 2, receptor 43
- Gene Aliases
-
EC2-V2R, Gm1961
- Chromosome Location
-
Chr.7: 8244348 - 8260599 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198961.2 |
NP_945199.1 |
4 - 5 |
1520 |
98 |
GenBank mRNA |
AJ543405.1 |
- |
4 - 5 |
1522 |
98 |
- Gene Symbol
- Vmn2r44
- Entrez Gene ID
-
434113
- Gene Name
- vomeronasal 2, receptor 44
- Gene Aliases
-
EG434113
- Chromosome Location
-
Chr.7: 8367460 - 8383238 on Build GRCm38
View Full Product Details